Assay Details
| Gene Symbol: LOC4339143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782297.1 | XP_015637783.1 | 1 - 2 | 174 | 97 | |
| GenBank mRNA | AK121022.1 | - | 1 - 2 | 153 | 97 | |
| BT131037.1 | - | 1 - 2 | 82 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015782297.1 transcripts.
Set Membership: |
|

LOC4339143
Genomic Map