Assay Details
Gene Symbol: LOC4339143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782297.1 | XP_015637783.1 | 4 - 5 | 451 | 76 | |
GenBank mRNA | AK121022.1 | - | 4 - 5 | 430 | 76 | |
BT131037.1 | - | 4 - 5 | 359 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
