Assay Details
Gene Symbol: LOC9267154 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015782485.2 | XP_015637971.1 | 19 - 20 | 2922 | 142 | |
GenBank mRNA | AK064293.1 | - | 4 - 5 | 548 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
