Assay Details
Gene Symbol: LOC4339268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784027.2 | XP_015639513.1 | 5 - 6 | 690 | 92 | |
GenBank mRNA | AK101520.1 | - | 5 - 6 | 648 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
