Assay Details
Gene Symbol: LOC4339268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015784027.2 | XP_015639513.1 | 9 - 10 | 1448 | 121 | |
GenBank mRNA | AK063235.1 | - | 2 - 3 | 468 | 121 | |
AK101520.1 | - | 9 - 10 | 1406 | 121 | ||
AK105718.1 | - | 2 - 3 | 191 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
