Assay Details
| Gene Symbol: LOC4339765 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015781969.2 | XP_015637455.1 | 1 - 2 | 1219 | 144 | |
| GenBank mRNA | AK120811.1 | - | 1 - 2 | 1183 | 144 | |
More Information
Set Membership: |
|

LOC4339765
Genomic Map