Assay Details
| Gene Symbol: LOC4339802 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783736.1 | XP_015639222.1 | 22 - 23 | 5993 | 78 | |
| XM_015783739.1 | XP_015639225.1 | 22 - 23 | 5993 | 78 | ||
| XM_015783741.1 | XP_015639227.1 | 22 - 23 | 5990 | 78 | ||
| XM_015783742.1 | XP_015639228.1 | 23 - 24 | 5912 | 78 | ||
| GenBank mRNA | AK106591.1 | - | 8 - 9 | 2289 | 79 | |
More Information
Set Membership: |
|

LOC4339802
Genomic Map