Assay Details
Gene Symbol: LOC4340169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787704.2 | XP_015643190.1 | 2 - 3 | 436 | 72 | |
XM_015787705.2 | XP_015643191.1 | 2 - 3 | 439 | 72 | ||
XM_015787706.2 | XP_015643192.1 | 2 - 3 | 369 | 72 | ||
GenBank mRNA | AK067799.1 | - | 1 - 2 | 585 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
