Assay Details
Gene Symbol: LOC4340169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787704.2 | XP_015643190.1 | 10 - 11 | 1386 | 69 | |
XM_015787706.2 | XP_015643192.1 | 10 - 11 | 1319 | 69 | ||
GenBank mRNA | AK067799.1 | - | 5 - 6 | 2086 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
