Assay Details
| Gene Symbol: LOC4340563 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786302.2 | XP_015641788.1 | 2 | 1833 | 113 | |
| GenBank mRNA | AK063733.1 | - | 1 | 52 | 113 | |
More Information
Set Membership: |
|

LOC4340563
Genomic Map