Assay Details
Gene Symbol: LOC4342077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787016.1 | XP_015642502.1 | 13 - 14 | 1914 | 69 | |
GenBank mRNA | AK062023.1 | - | 8 - 9 | 1185 | 69 | |
AK102478.1 | - | 13 - 14 | 1863 | 69 | ||
AK119605.1 | - | 1103 | 69 | |||
GU120345.1 | - | 13 - 14 | 1840 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
