Assay Details
| Gene Symbol: LOC4342077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787016.1 | XP_015642502.1 | 14 - 15 | 2319 | 64 | |
| GenBank mRNA | AK062023.1 | - | 9 - 10 | 1590 | 64 | |
| AK102478.1 | - | 14 - 15 | 2268 | 64 | ||
| AK119605.1 | - | 1508 | 64 | |||
| GU120345.1 | - | 14 - 15 | 2245 | 64 | ||
More Information
Set Membership: |
|

LOC4342077
Genomic Map