Assay Details
| Gene Symbol: LOC4342077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787016.1 | XP_015642502.1 | 6 - 7 | 796 | 68 | |
| GenBank mRNA | AK062023.1 | - | 1 - 2 | 67 | 68 | |
| AK063385.1 | - | 6 - 7 | 741 | 68 | ||
| AK102478.1 | - | 6 - 7 | 745 | 68 | ||
| GU120345.1 | - | 6 - 7 | 722 | 68 | ||
More Information
Set Membership: |
|

LOC4342077
Genomic Map