Assay Details
| Gene Symbol: LOC4342077 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787016.1 | XP_015642502.1 | 8 - 9 | 1067 | 97 | |
| GenBank mRNA | AK062023.1 | - | 3 - 4 | 338 | 97 | |
| AK102478.1 | - | 8 - 9 | 1016 | 97 | ||
| GU120345.1 | - | 8 - 9 | 993 | 97 | ||
More Information
Set Membership: |
|

LOC4342077
Genomic Map