Assay Details
Gene Symbol: LOC4342129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787786.2 | XP_015643272.1 | 2 - 3 | 217 | 84 | |
GenBank mRNA | AK062226.1 | - | 2 - 3 | 193 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015787786.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
