Assay Details
| Gene Symbol: LOC4342129 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787785.2 | XP_015643271.1 | 3 - 4 | 341 | 88 | |
| XM_015787786.2 | XP_015643272.1 | 3 - 4 | 333 | 88 | ||
| GenBank mRNA | AF093632.1 | - | 3 - 4 | 236 | 88 | |
| AK062226.1 | - | 3 - 4 | 309 | 88 | ||
| AK068898.1 | - | 2 - 3 | 88 | |||
| BT129146.1 | - | 3 - 4 | 305 | 88 | ||
| BT130172.1 | - | 3 - 4 | 295 | 88 | ||
| BT131862.1 | - | 1 - 2 | 142 | 88 | ||
| EU847026.1 | - | 3 - 4 | 262 | 88 | ||
More Information
Set Membership: |
|

LOC4342129
Genomic Map