Assay Details
Gene Symbol: LOC4342129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015787785.2 | XP_015643271.1 | 5 - 6 | 441 | 118 | |
XM_015787786.2 | XP_015643272.1 | 5 - 6 | 433 | 118 | ||
GenBank mRNA | AF093632.1 | - | 5 - 6 | 336 | 118 | |
AK062226.1 | - | 5 - 6 | 409 | 118 | ||
AK068898.1 | - | 4 - 5 | 494 | 118 | ||
BT129146.1 | - | 5 - 6 | 405 | 118 | ||
BT129798.1 | - | 2 - 3 | 68 | 118 | ||
BT130172.1 | - | 5 - 6 | 395 | 118 | ||
BT131862.1 | - | 3 - 4 | 242 | 118 | ||
EU847026.1 | - | 5 - 6 | 362 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
