Assay Details
Gene Symbol: LOC4343098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789128.2 | XP_015644614.1 | 1 - 2 | 651 | 80 | |
GenBank mRNA | AK073296.1 | - | 1 - 2 | 346 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
