Assay Details
| Gene Symbol: LOC4343276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791756.2 | XP_015647242.1 | 23 - 24 | 2960 | 82 | |
| XM_015791757.2 | XP_015647243.1 | 23 - 24 | 2962 | 82 | ||
| XM_015791758.2 | XP_015647244.1 | 23 - 24 | 2902 | 82 | ||
| XM_015791760.2 | XP_015647246.1 | 23 - 24 | 2960 | 82 | ||
| XM_015791761.2 | XP_015647247.1 | 23 - 24 | 2958 | 82 | ||
| XM_015791762.2 | XP_015647248.1 | 23 - 24 | 2959 | 82 | ||
| XM_015791763.2 | XP_015647249.1 | 23 - 24 | 2914 | 82 | ||
| GenBank mRNA | AK067461.1 | - | 23 - 24 | 2919 | 82 | |
More Information
Set Membership: |
|

LOC4343276
Genomic Map