Assay Details
| Gene Symbol: LOC4343276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791756.2 | XP_015647242.1 | 24 - 25 | 3020 | 65 | |
| XM_015791758.2 | XP_015647244.1 | 24 - 25 | 2962 | 65 | ||
| XM_015791760.2 | XP_015647246.1 | 24 - 25 | 3020 | 65 | ||
| XM_015791761.2 | XP_015647247.1 | 24 - 25 | 3018 | 65 | ||
| XM_015791762.2 | XP_015647248.1 | 24 - 25 | 3019 | 65 | ||
| XM_015791763.2 | XP_015647249.1 | 24 - 25 | 2974 | 65 | ||
| XM_015791764.2 | XP_015647250.1 | 24 - 25 | 3028 | 65 | ||
| GenBank mRNA | AK067461.1 | - | 24 - 25 | 2979 | 65 | |
More Information
Set Membership: |
|

LOC4343276
Genomic Map