Assay Details
| Gene Symbol: LOC4343276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791756.2 | XP_015647242.1 | 25 - 26 | 3179 | 131 | |
| XM_015791757.2 | XP_015647243.1 | 25 - 26 | 3175 | 131 | ||
| XM_015791758.2 | XP_015647244.1 | 25 - 26 | 3121 | 131 | ||
| XM_015791759.2 | XP_015647245.1 | 24 - 25 | 3124 | 131 | ||
| XM_015791760.2 | XP_015647246.1 | 25 - 26 | 3179 | 131 | ||
| XM_015791761.2 | XP_015647247.1 | 25 - 26 | 3177 | 131 | ||
| XM_015791762.2 | XP_015647248.1 | 25 - 26 | 3178 | 131 | ||
| XM_015791763.2 | XP_015647249.1 | 25 - 26 | 3133 | 131 | ||
| GenBank mRNA | AK067461.1 | - | 25 - 26 | 3138 | 131 | |
More Information
Set Membership: |
|

LOC4343276
Genomic Map