Assay Details
| Gene Symbol: LOC4343276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791756.2 | XP_015647242.1 | 29 - 30 | 3525 | 67 | |
| XM_015791757.2 | XP_015647243.1 | 29 - 30 | 3521 | 67 | ||
| XM_015791758.2 | XP_015647244.1 | 29 - 30 | 3467 | 67 | ||
| XM_015791759.2 | XP_015647245.1 | 28 - 29 | 3470 | 67 | ||
| XM_015791760.2 | XP_015647246.1 | 29 - 30 | 3447 | 67 | ||
| XM_015791761.2 | XP_015647247.1 | 28 - 29 | 3388 | 67 | ||
| XM_015791762.2 | XP_015647248.1 | 27 - 28 | 3338 | 67 | ||
| XM_015791763.2 | XP_015647249.1 | 28 - 29 | 3344 | 67 | ||
| GenBank mRNA | AK067461.1 | - | 28 - 29 | 3349 | 67 | |
More Information
Set Membership: |
|

LOC4343276
Genomic Map