Assay Details
| Gene Symbol: LOC4343276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791756.2 | XP_015647242.1 | 30 - 31 | 3581 | 66 | |
| XM_015791757.2 | XP_015647243.1 | 30 - 31 | 3577 | 66 | ||
| XM_015791758.2 | XP_015647244.1 | 30 - 31 | 3523 | 66 | ||
| XM_015791759.2 | XP_015647245.1 | 29 - 30 | 3526 | 66 | ||
| XM_015791760.2 | XP_015647246.1 | 30 - 31 | 3503 | 66 | ||
| XM_015791761.2 | XP_015647247.1 | 29 - 30 | 3444 | 66 | ||
| XM_015791762.2 | XP_015647248.1 | 28 - 29 | 3394 | 66 | ||
| XM_015791763.2 | XP_015647249.1 | 29 - 30 | 3400 | 66 | ||
| GenBank mRNA | AK067461.1 | - | 29 - 30 | 3405 | 66 | |
More Information
Set Membership: |
|

LOC4343276
Genomic Map