Assay Details
| Gene Symbol: LOC4343276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791758.2 | XP_015647244.1 | 2 - 3 | 614 | 82 | |
| XM_015791763.2 | XP_015647249.1 | 2 - 3 | 626 | 82 | ||
| GenBank mRNA | AK067461.1 | - | 2 - 3 | 632 | 82 | |
More Information
Set Membership: |
|

LOC4343276
Genomic Map