Assay Details
| Gene Symbol: LOC4343276 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791756.2 | XP_015647242.1 | 6 - 7 | 1245 | 86 | |
| XM_015791757.2 | XP_015647243.1 | 6 - 7 | 1247 | 86 | ||
| XM_015791758.2 | XP_015647244.1 | 6 - 7 | 1187 | 86 | ||
| XM_015791759.2 | XP_015647245.1 | 6 - 7 | 1247 | 86 | ||
| XM_015791760.2 | XP_015647246.1 | 6 - 7 | 1245 | 86 | ||
| XM_015791761.2 | XP_015647247.1 | 6 - 7 | 1243 | 86 | ||
| XM_015791762.2 | XP_015647248.1 | 6 - 7 | 1244 | 86 | ||
| XM_015791763.2 | XP_015647249.1 | 6 - 7 | 1199 | 86 | ||
| XM_015791764.2 | XP_015647250.1 | 6 - 7 | 1249 | 86 | ||
| XM_015791765.2 | XP_015647251.1 | 6 - 7 | 1249 | 86 | ||
| GenBank mRNA | AK067461.1 | - | 6 - 7 | 1204 | 86 | |
More Information
Set Membership: |
|

LOC4343276
Genomic Map