Assay Details
| Gene Symbol: LOC4343413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791090.2 | XP_015646576.1 | 1 - 2 | 974 | 77 | |
| GenBank mRNA | AK067405.1 | - | 2 - 3 | 821 | 77 | |
| AK103813.1 | - | 1 - 2 | 966 | 77 | ||
More Information
Set Membership: |
|

LOC4343413
Genomic Map