Assay Details
| Gene Symbol: LOC4343413 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791090.2 | XP_015646576.1 | 5 - 6 | 3037 | 78 | |
| GenBank mRNA | AK067405.1 | - | 6 - 7 | 2902 | 78 | |
More Information
Set Membership: |
|

LOC4343413
Genomic Map