Assay Details
Gene Symbol: LOC4344157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792522.2 | XP_015648008.1 | 3 - 4 | 568 | 78 | |
XM_015792523.2 | XP_015648009.1 | 3 - 4 | 562 | 78 | ||
XM_015792524.2 | XP_015648010.1 | 3 - 4 | 1058 | 78 | ||
GenBank mRNA | AK058476.1 | - | 3 - 4 | 462 | 78 | |
AK102906.1 | - | 3 - 4 | 978 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
