Assay Details
| Gene Symbol: LOC4344157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792523.2 | XP_015648009.1 | 4 - 5 | 684 | 67 | |
| XM_015792524.2 | XP_015648010.1 | 4 - 5 | 1180 | 67 | ||
| GenBank mRNA | AK058476.1 | - | 4 - 5 | 584 | 67 | |
| AK102906.1 | - | 4 - 5 | 1100 | 67 | ||
More Information
Set Membership: |
|

LOC4344157
Genomic Map