Assay Details
| Gene Symbol: LOC4344163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792552.2 | XP_015648038.1 | 4 - 5 | 831 | 117 | |
| XM_015792553.1 | XP_015648039.1 | 4 - 5 | 544 | 117 | ||
| GenBank mRNA | AK103870.1 | - | 4 - 5 | 545 | 117 | |
More Information
Set Membership: |
|

LOC4344163
Genomic Map