Assay Details
| Gene Symbol: LOC9271247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794773.2 | XP_015650259.1 | 8 - 9 | 1208 | 105 | |
| GenBank mRNA | AK101379.1 | - | 5 - 6 | 2426 | 105 | |
More Information
Set Membership: |
|

LOC9271247
Genomic Map