Assay Details
| Gene Symbol: LOC4345339 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795484.1 | XP_015650970.1 | 7 - 8 | 1435 | 82 | |
| XM_015795485.2 | XP_015650971.1 | 7 - 8 | 1331 | 82 | ||
| GenBank mRNA | AK058246.1 | - | 1 - 2 | 94 | 82 | |
| AK071826.1 | - | 7 - 8 | 1444 | 82 | ||
| AK103906.1 | - | 7 - 8 | 1315 | 82 | ||
| D83539.1 | - | 7 - 8 | 1288 | 82 | ||
| EF122437.2 | - | 7 - 8 | 1300 | 82 | ||
| FJ940194.1 | - | 7 - 8 | 1351 | 82 | ||
More Information
Set Membership: |
|

LOC4345339
Genomic Map