Assay Details
Gene Symbol: LOC4345459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795350.2 | XP_015650836.1 | 1 - 2 | 439 | 89 | |
XM_015795352.2 | XP_015650838.1 | 1 - 2 | 461 | 89 | ||
GenBank mRNA | AK073889.1 | - | 1 - 2 | 431 | 89 | |
AK108397.1 | - | 1 - 2 | 429 | 89 | ||
AK119576.1 | - | 1 - 2 | 466 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
