Assay Details
Gene Symbol: LOC4345459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795350.2 | XP_015650836.1 | 4 - 5 | 702 | 67 | |
XM_015795352.2 | XP_015650838.1 | 4 - 5 | 724 | 67 | ||
GenBank mRNA | AK073889.1 | - | 4 - 5 | 694 | 67 | |
AK119576.1 | - | 4 - 5 | 729 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
