Assay Details
Gene Symbol: LOC4345459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795350.2 | XP_015650836.1 | 5 - 6 | 867 | 94 | |
XM_015795352.2 | XP_015650838.1 | 5 - 6 | 889 | 94 | ||
GenBank mRNA | AK073889.1 | - | 5 - 6 | 859 | 94 | |
AK119576.1 | - | 5 - 6 | 894 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
