Assay Details
Gene Symbol: LOC4345459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795350.2 | XP_015650836.1 | 6 - 7 | 965 | 56 | |
XM_015795352.2 | XP_015650838.1 | 6 - 7 | 987 | 56 | ||
GenBank mRNA | AK073889.1 | - | 6 - 7 | 957 | 56 | |
AK119576.1 | - | 6 - 7 | 992 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
