Assay Details
Gene Symbol: LOC4345459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795350.2 | XP_015650836.1 | 9 - 10 | 1264 | 66 | |
XM_015795352.2 | XP_015650838.1 | 9 - 10 | 1286 | 66 | ||
GenBank mRNA | AK073889.1 | - | 9 - 10 | 1256 | 66 | |
AK119576.1 | - | 9 - 10 | 1291 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
