Assay Details
Gene Symbol: LOC4341799 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015788238.2 | XP_015643724.1 | 5 - 6 | 879 | 91 | |
XM_015788239.2 | XP_015643725.1 | 5 - 6 | 740 | 91 | ||
GenBank mRNA | AK070871.1 | - | 4 - 5 | 473 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
