Assay Details
| Gene Symbol: LOC4342497 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792191.2 | XP_015647677.1 | 8 - 9 | 947 | 125 | |
| GenBank mRNA | AK065622.1 | - | 8 - 9 | 943 | 125 | |
More Information
Set Membership: |
|

LOC4342497
Genomic Map