Assay Details
| Gene Symbol: LOC4343971 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789227.2 | XP_015644713.1 | 4 - 5 | 717 | 103 | |
| GenBank mRNA | AK288055.1 | - | 4 - 5 | 691 | 103 | |
More Information
Set Membership: |
|

LOC4343971
Genomic Map