Assay Details
Gene Symbol: LOC4344036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015790645.2 | XP_015646131.2 | 7 - 8 | 786 | 127 | |
GenBank mRNA | AK107137.1 | - | 7 - 8 | 791 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
