Assay Details
Gene Symbol: LOC4344046 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789701.2 | XP_015645187.1 | 1 - 2 | 237 | 104 | |
GenBank mRNA | AK073273.1 | - | 1 - 2 | 225 | 104 | |
AK104089.1 | - | 1 - 2 | 224 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
