Assay Details
Gene Symbol: LOC4344840 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015794716.2 | XP_015650202.1 | 1 - 2 | 183 | 79 | |
GenBank mRNA | AF032971.1 | - | 1 - 2 | 149 | 79 | |
AK059338.1 | - | 1 - 2 | 175 | 79 | ||
BT129153.1 | - | 1 - 2 | 161 | 79 | ||
BT131332.1 | - | 1 - 2 | 174 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
