Assay Details
Gene Symbol: LOC9269126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792660.2 | XP_015648146.1 | 4 - 5 | 597 | 62 | |
GenBank mRNA | AK240826.1 | - | 4 - 5 | 601 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
