Assay Details
Gene Symbol: LOC9269126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015792660.2 | XP_015648146.1 | 5 - 6 | 741 | 102 | |
GenBank mRNA | AK240826.1 | - | 5 - 6 | 745 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
