Assay Details
| Gene Symbol: LOC4345051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792922.2 | XP_015648408.1 | 2 - 3 | 324 | 99 | |
| GenBank mRNA | AK103017.1 | - | 2 - 3 | 331 | 99 | |
More Information
Set Membership: |
|

LOC4345051
Genomic Map