Assay Details
| Gene Symbol: LOC4346300 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792811.2 | XP_015648297.1 | 2 - 3 | 542 | 69 | |
| GenBank mRNA | AK061111.1 | - | 2 - 3 | 518 | 69 | |
| AY345242.1 | - | 2 - 3 | 195 | 69 | ||
| AY581257.1 | - | 2 - 3 | 286 | 69 | ||
More Information
Set Membership: |
|

LOC4346300
Genomic Map