Assay Details
Gene Symbol: LOC4346428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756794.2 | XP_015612280.1 | 1 - 2 | 404 | 137 | |
GenBank mRNA | AK103665.1 | - | 1 - 2 | 394 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() |
