Assay Details
Gene Symbol: LOC4346428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015756794.2 | XP_015612280.1 | 4 - 5 | 752 | 142 | |
GenBank mRNA | AK103665.1 | - | 4 - 5 | 742 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
