Assay Details
Gene Symbol: LOC4346747 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015795965.2 | XP_015651451.1 | 1 - 2 | 156 | 90 | |
GenBank mRNA | AK068681.1 | - | 1 - 2 | 111 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
